Thiophene-2-methylamine 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A11700-22
A11700-06
A11700-14
CAAAAA11700-22EA
736.28
CAD
CAAAAA11700-22
CAAAA11700-06
CAAAA11700-14
Thiophene-2-methylamine 97%
Thiophene-2-methylamine
Purity: 97%
CAS No.: 27757-85-3
Molecular Formula: C5H7NS
Formula Weight: 113.19
Flash Point: 73°(163°F)
Refractive Index: 1.5670
Boiling Point: 85-88°/14mm
Beilstein Reference: 106296
UN No.: 2735
MDL No.: MFCD00005460
Formula:
C₅H₇NS Boiling Pt: 85…88 °C/14 mm Density: 1.100 Flash Pt: 73 °C(163 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00005460 CAS Number: 27757-85-3 EINECS: 248-639-6 UN: 2735 ADR: "8",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...