5-Acetylsalicylic acid 98+%

Supplier: Thermo Scientific Chemicals
Warning

A11663-22 A11663-06 A11663-14
CAAAAA11663-22EA 1464.66 CAD
CAAAAA11663-22 CAAAA11663-06 CAAAA11663-14
5-Acetylsalicylic acid 98+%
5-Acetylsalicylic acid

Purity: 98+% CAS No.: 13110-96-8 Molecular Formula: C9H8O4 Formula Weight: 180.15 Flash Point: 250°(482°F) Melting Point: 214-216° Beilstein Reference: 2096968 MDL No.: MFCD00013978

Formula: C₉H₈O₄
Melting Pt: 214…216 °C
Flash Pt: 250 °C(482 °F)
Storage Temperature: Ambient
MDL Number: MFCD00013978
CAS Number: 13110-96-8
EINECS: 236-037-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR