9-Ethylcarbazole 99%
Supplier: Thermo Scientific Chemicals
A11653-22
A11653-0B
A11653-30
CAAAAA11653-22EA
75.05
CAD
CAAAAA11653-22
CAAAA11653-0B
CAAAA11653-30
9-Ethylcarbazole 99%
9-Ethylcarbazole
Purity: 99%
CAS No.: 86-28-2
Molecular Formula: C14H13N
Formula Weight: 195.27
Flash Point: 186°(367°F)
Melting Point: 67-71°
Boiling Point: 166°/4mm
Beilstein Reference: 148115
MDL No.: MFCD00004967
Formula:
C₁₄H₁₃N Boiling Pt: 166 °C/4 mm Melting Pt: 67…71 °C Flash Pt: 186 °C(367 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00004967 CAS Number: 86-28-2 EINECS: 201-660-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...