1-Dimethylamino-2-nitroethylene 98%

Supplier: Thermo Scientific Chemicals
Danger

A11637-03 A11637-06
CAAAA11637-03EA 55.62 CAD
CAAAA11637-03 CAAAA11637-06
1-Dimethylamino-2-nitroethylene 98%
1-Dimethylamino-2-nitroethylene

Purity: 98% CAS No.: 1190-92-7 Molecular Formula: (CH3)2NCH=CHNO2 Formula Weight: 116.12 Storage Temperature: 2°C to 8°C Melting Point: 101-105° Beilstein Reference: 1903136 MDL No.: MFCD00051519

Formula: C₄H₈N₂O₂
MW: 116.12 g/mol
Melting Pt: 101…105 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00051519
CAS Number: 1190-92-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR