N-Acetyl-L-tyrosine ethyl ester monohydrate 97%
Supplier: Thermo Scientific Chemicals
A11577-06
A11577-14
CAAAA11577-06EA
85.16
CAD
CAAAA11577-06
CAAAA11577-14
N-Acetyl-L-tyrosine ethyl ester monohydrate 97%
N-Acetyl-L-tyrosine ethyl ester monohydrate
Purity: 97%
CAS No.: 36546-50-6
Molecular Formula: C13H17NO4.H2O
Formula Weight: 269.30 (251.28anhy)
Storage Temperature: -30°C to -10°C
Melting Point: 80-81°
MDL No.: MFCD00149093
Formula:
C₁₃H₁₉NO₅ Melting Pt: 80…81 °C Storage Temperature: Freezer |
MDL Number:
MFCD00149093 CAS Number: 36546-50-6 EINECS: 212-663-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...