Sodium dodecatungstophosphate hydrate

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Phosphotungstic acid sodium salt hydrate

A11575-14 A11575-22 A11575-36
CAAAAA11575-14EA 109.81 CAD
CAAAAA11575-14 CAAAAA11575-22 CAAAAA11575-36
Sodium dodecatungstophosphate hydrate
Phosphotungstic acid sodium salt hydrate

CAS No.: 51312-42-6 Molecular Formula: Na3PO4.12WO3.xH2O Formula Weight: 2946.12(anhy) Melting Point: 350° MDL No.: MFCD00151658

Formula: Na₃O₄₀PW₁₂·H₂O
MW: 2946.12 g/mol (anhy)
Melting Pt: 350 °C
Storage Temperature: Ambient
MDL Number: MFCD00151658
CAS Number: 51312-42-6
EINECS: 257-132-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR