Diethyl (2-cyanoethyl)malonate 98+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Diethyl 2-(2-cyanoethyl)malonate

A11567-14 A11567-09 A11567-18
CAAAAA11567-14EA 124.35 CAD
CAAAAA11567-14 CAAAA11567-09 CAAAA11567-18
Diethyl (2-cyanoethyl)malonate 98+%
Diethyl (2-cyanoethyl)malonate

Purity: 98+% CAS No.: 17216-62-5 Molecular Formula: C10H15NO4 Formula Weight: 213.23 Flash Point: 110°(230°F) Refractive Index: 1.4360 Boiling Point: 115-117°/2mm Beilstein Reference: 1790237 UN No.: 3276 MDL No.: MFCD00001966

Formula: C₁₀H₁₅NO₄
MW: 213.23 g/mol
Boiling Pt: 115…117 °C (2 mmHg)
Density: 1.079
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001966
CAS Number: 17216-62-5
EINECS: 241-260-7
UN: 2810
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR