1-Ethyl-3-methyl-1H-pyrazole-5-carboxylic acid 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
1-Ethyl-3-methylpyrazole-5-carboxylic acid
A11561-03
A11561-06
CAAAA11561-03EA
113.29
CAD
CAAAA11561-03
CAAAA11561-06
1-Ethyl-3-methyl-1H-pyrazole-5-carboxylic acid 97%
1-Ethyl-3-methyl-1H-pyrazole-5-carboxylic acid
Purity: 97%
CAS No.: 50920-65-5
Molecular Formula: C7H10N2O2
Formula Weight: 154.17
Melting Point: 137-140°
MDL No.: MFCD00156147
Formula:
C₇H₁₀N₂O₂ MW: 154.17 g/mol Melting Pt: 137…140 °C Storage Temperature: Ambient |
MDL Number:
MFCD00156147 CAS Number: 50920-65-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...