Methyl diphenylphosphinite 98+%

Supplier: Thermo Scientific Chemicals
Warning

A11555-03 A11555-14
CAAAAA11555-03EA 124.82 CAD
CAAAAA11555-03 CAAAA11555-14
Methyl diphenylphosphinite 98+%
Methyl diphenylphosphinite

Purity: 98+% CAS No.: 4020-99-9 Molecular Formula: C13H13OP Formula Weight: 216.22 Storage Temperature: -30°C to -10°C Flash Point: 110°(230°F) Refractive Index: 1.6040 Boiling Point: 148-150°/6mm Beilstein Reference: 1640480 MDL No.: MFCD00048025

Formula: C₁₃H₁₃OP
Boiling Pt: 148…150 °C/6 mm
Density: 1,078
Flash Pt: >110 °C(230 °F)
Storage Temperature: Freezer
MDL Number: MFCD00048025
CAS Number: 4020-99-9
EINECS: 223-683-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR