Ethyl-5-amino-1-methyl-1H-pyrazole-4-carboxylate 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A11534-06
A11534-14
CAAAAA11534-22EA
2341.88
CAD
CAAAAA11534-22
CAAAA11534-06
CAAAA11534-14
Ethyl-5-amino-1-methyl-1H-pyrazole-4-carboxylate 98%
Ethyl 5-amino-1-methyl-1H-pyrazole-4-carboxylate
Purity: 98%
CAS No.: 31037-02-2
Molecular Formula: C7H11N3O2
Formula Weight: 169.18
Melting Point: 97-102°
Beilstein Reference: 131200
Formula:
C₇H₁₁N₃O₂ Melting Pt: 97…102 °C Storage Temperature: Ambient |
MDL Number:
MFCD00051652 CAS Number: 31037-02-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...