Methyl-ɑ-D-mannopyranoside 99%
Supplier: Thermo Scientific Chemicals
Synonyms:
ɑ-Methyl-D-mannoside, Methyl-ɑ-D-Mannoside, Methyl α-D-Mannopyranoside, ɑ-Methyl-D-mannopyranoside, (2R,3S,4S,5S,6S)-2-(Hydroxymethyl)-6-methoxytetrahydro-2H-pyran-3,4,5-triol
A11533-14
A11533-22
CAAAA11533-14EA
83.27
CAD
CAAAA11533-14
CAAAA11533-22
Methyl-ɑ-D-mannopyranoside 99%
Methyl-alpha-D-mannopyranoside
Purity: 99%
CAS No.: 617-04-9
Molecular Formula: C7H14O6
Formula Weight: 194.18
Melting Point: 193-196°
Beilstein Reference: 81566
Optical Rotation: +79° (c=10 in water)
MDL No.: MFCD00063262
Formula:
C₇H₁₄O₆ MW: 194.18 g/mol Melting Pt: 193…196 °C Storage Temperature: Ambient |
MDL Number:
MFCD00063262 CAS Number: 617-04-9 EINECS: 210-502-3 |
Specification Test Results
Optical rotation | +79° (c=4 in water) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...