Methyl p-Tolyl sulfone 99%

Supplier: Thermo Scientific Chemicals

Synonyms: 1-Methyl-4-(methylsulfonyl)benzene, 4-(Methylsulfonyl)toluene, 4-(Methylsulphonyl)toluene, 1-Methyl-4-(methylsulphonyl)benzene

A11531-06 A11531-14
CAAAA11531-06EA 73.63 CAD
CAAAA11531-06 CAAAA11531-14 CAAAA11531-22
Methyl p-Tolyl sulfone 99%
Methyl p-Tolyl sulfone

Purity: 99% CAS No.: 3185-99-7 Molecular Formula: C8H10O2S Formula Weight: 170.23 Melting Point: 86-89° Boiling Point: 140°/3mm Beilstein Reference: 607296 MDL No.: MFCD00014742

Formula: C₈H₁₀O₂S
MW: 170.23 g/mol
Boiling Pt: 140 °C/3 mm
Melting Pt: 86…89 °C
Storage Temperature: Ambient
MDL Number: MFCD00014742
CAS Number: 3185-99-7
EINECS: 221-682-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR