Salicylohydroxamic acid 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A11528-14
A11528-22
A11528-09
A11528-18
CAAAAA11528-14EA
112.02
CAD
CAAAAA11528-14
CAAAAA11528-22
CAAAA11528-09
CAAAA11528-18
Salicylohydroxamic acid 99%
Salicylohydroxamic acid
Purity: 99%
CAS No.: 89-73-6
Molecular Formula: C7H7NO3
Formula Weight: 153.14
Melting Point: ca 170° dec.
Beilstein Reference: 1210520
Merck Reference: 14,8331
MDL No.: MFCD00002110
Formula:
C₇H₇NO₃ Melting Pt: ca 170 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00002110 CAS Number: 89-73-6 EINECS: 201-934-3 Merck Index: 14,8331 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...