2-Chlorophenyl phosphorodichloridate 98+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A11505-06
A11505-14
A11505-22
CAAAA11505-06EA
49.93
CAD
CAAAA11505-06
CAAAA11505-14
CAAAA11505-22
2-Chlorophenyl phosphorodichloridate 98+%
2-Chlorophenyl phosphorodichloridate
Purity: 98+%
CAS No.: 15074-54-1
Molecular Formula: C6H4Cl3O2P
Formula Weight: 245.43
Flash Point: 105°(221°F)
Refractive Index: 1.5380
Boiling Point: 82-84°/0.4mm
Beilstein Reference: 911009
UN No.: 3265
MDL No.: MFCD00009704
Formula:
C₆H₄Cl₃O₂P MW: 245.43 g/mol Boiling Pt: 82…84 °C (0.4 mmHg) Density: 1.491 Flash Pt: 105 °C (221 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009704 CAS Number: 15074-54-1 EINECS: 239-129-4 UN: 3265 ADR: 8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...