Phenylfluorone
Supplier: Thermo Scientific Chemicals
A11495-06
A11495-14
A11495-22
CAAAA11495-06EA
87.53
CAD
CAAAA11495-06
CAAAA11495-14
CAAAA11495-22
Phenylfluorone
Phenylfluorone
CAS No.: 975-17-7
Molecular Formula: C19H12O5
Formula Weight: 320.30
Melting Point:
Formula:
C₁₉H₁₂O₅ Melting Pt: >300 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005048 CAS Number: 975-17-7 EINECS: 213-550-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...