ɑ-Chloralose mixture of anomers <20% β anomer ≥98%

Supplier: Thermo Scientific Chemicals
Danger

A11492-14 A11492-22
CAAAA11492-14EA 39.18 CAD
CAAAA11492-14 CAAAA11492-22
ɑ-Chloralose mixture of anomers <20% β anomer ≥98%
alpha-Chloralose

Used as sedative and anesthetic in lab animals


Purity: 98+% CAS No.: 15879-93-3 Molecular Formula: C8H11Cl3O6 Formula Weight: 309.53 Melting Point: 180-184° Beilstein Reference: 85418 Optical Rotation: +19° (c=2 in ethanol) MDL No.: MFCD00005542

Formula: C₈H₁₁Cl₃O₆
MW: 309.52 g/mol
Melting Pt: 180…184 °C
Storage Temperature: Ambient
MDL Number: MFCD00005542
CAS Number: 15879-93-3
EINECS: 240-016-7
Merck Index: 14,2072

Order Now

Specification Test Results

Optical rotation +19° (c=2 in ethanol)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR