1-Isopropyl-4-piperidone 99%

Supplier: Thermo Scientific Chemicals
Danger

A11480-09 A11480-18 A11480-30
CAAAA11480-09EA 50.4 CAD
CAAAA11480-09 CAAAA11480-18 CAAAA11480-30
1-Isopropyl-4-piperidone 99%
1-Isopropyl-4-piperidone

Purity: 99% CAS No.: 5355-68-0 Molecular Formula: C8H15NO Formula Weight: 141.22 Refractive Index: 1.4655 Boiling Point: 100-101°/27mm MDL No.: MFCD00038035

Formula: C₈H₁₅NO
MW: 141.22 g/mol
Boiling Pt: 100…101 °C/27 mm
Density: 0.950
Storage Temperature: Ambient
MDL Number: MFCD00038035
CAS Number: 5355-68-0
EINECS: 226-339-6
UN: 2735
ADR: "8",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR