Theophylline-7-acetic acid 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A11473-36
A11473-14
A11473-22
CAAAAA11473-36EA
2485.34
CAD
CAAAAA11473-36
CAAAA11473-14
CAAAA11473-22
Theophylline-7-acetic acid 98%
Theophylline-7-acetic acid
Purity: 98%
CAS No.: 652-37-9
Molecular Formula: C9H10N4O4
Formula Weight: 238.20
Melting Point: 268-272°
Beilstein Reference: 279221
Merck Reference: 14,24
MDL No.: MFCD00022832
Formula:
C₉H₁₀N₄O₄ Melting Pt: 268…272 °C Storage Temperature: Ambient |
MDL Number:
MFCD00022832 CAS Number: 652-37-9 EINECS: 211-490-2 Merck Index: 14,24 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...