Dibenzyl carbonate 98%

Supplier: Thermo Scientific Chemicals
Warning

A11464-09 A11464-18 A11464-30
CAAAA11464-09EA 164.32 CAD
CAAAA11464-09 CAAAA11464-18 CAAAA11464-30
Dibenzyl carbonate 98%
Dibenzyl carbonate

Fieser: 19,117


Purity: 98% CAS No.: 3459-92-5 Molecular Formula: C15H14O3 Formula Weight: 242.27 Flash Point: 110°(230°F) Refractive Index: 1.5485 Melting Point: 27-30° Boiling Point: 150-155°/0.8mm Beilstein Reference: 1882864 MDL No.: MFCD00014436

Formula: C₁₅H₁₄O₃
MW: 242.27 g/mol
Boiling Pt: 150…155 °C (0.8 mmHg)
Melting Pt: 27…30 °C
Flash Pt: >110 °C (230 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00014436
CAS Number: 3459-92-5
EINECS: 222-401-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR