(S)-Methyl-2-amino-3-phenylpropanoate hydrochloride 98%

Supplier: Thermo Scientific Chemicals

Synonyms: L-Methyl phenylalanate hydrochloride, L-Phenylalanine methyl ester hydrochloride, H-Phe-OMe·HCl

A11455-09 A11455-18
CAAAA11455-09EA 78.37 CAD
CAAAA11455-09 CAAAA11455-18 CAAAA11455-30
(S)-Methyl-2-amino-3-phenylpropanoate hydrochloride 98%
(S)-Methyl-2-amino-3-phenylpropanoate hydrochloride

Purity: 99% CAS No.: 7524-50-7 Molecular Formula: C10H13NO2.HCl Formula Weight: 215.68 Melting Point: 158-162° Beilstein Reference: 3597948 Optical Rotation: +37° (c=2 in ethanol) MDL No.: MFCD00012489

Formula: C₁₀H₁₄ClNO₂
MW: 215.68 g/mol
Melting Pt: 158…162 °C
Storage Temperature: Ambient
MDL Number: MFCD00012489
CAS Number: 7524-50-7
EINECS: 231-383-4

Order Now

Specification Test Results

Optical rotation +37° (c=2 in ethanol)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR