D-(-)-Penicillamine 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
H-D-Pen-OH, (S)-2-Amino-3-mercapto-3-methylbutanoic acid, Penicillamine
Exogenous nitric oxide synthase (NOS) modulator. Physiological chelating agent for heavy metals
Purity: 99%
CAS No.: 52-67-5
Molecular Formula: C5H11NO2S
Formula Weight: 149.21
Storage Temperature: 2°C to 8°C
Odor: Unpleasant
pH: 4.5-5.5
Melting Point: 202-204° dec.
Beilstein Reference: 1722375
Merck Reference: 14,7088
Optical Rotation: -63° (c=1 in 1N NaOH)
MDL No.: MFCD00064302
Formula:
C₅H₁₁NO₂S Melting Pt: 202…204 °C (dec.) Storage Temperature: Refrigerator |
MDL Number:
MFCD00064302 CAS Number: 52-67-5 EINECS: 200-148-8 Merck Index: 14,7088 |
Specification Test Results
Optical rotation | -63° (c=1 in 1N NaOH) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...