3-Amino-5-hydroxy-1H-pyrazole 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-Amino-5-hydroxy-1H-pyrazole, 3-Amino-5-hydroxypyrazole, 5-Amino-3-pyrazolol, 3-Amino-1H-pyrazol-5(4H)-one, 3-Amino-2-pyrazolin-5-one, 5-Hydroxy-3-aminopyrazole, 5-Amino-2,4-dihydro-3H-pyrazol-3-one

A11412-06 A11412-14 A11412-22
CAAAA11412-06EA 41.87 CAD
CAAAA11412-06 CAAAA11412-14 CAAAA11412-22
3-Amino-5-hydroxy-1H-pyrazole 98%
3-Amino-5-hydroxy-1H-pyrazole

Purity: 98% CAS No.: 6126-22-3 Molecular Formula: C3H5N3O Formula Weight: 99.09 Odor: Odorless Melting Point: ca 210° dec. Beilstein Reference: 507793 MDL No.: MFCD00022384

Formula: C₃H₅N₃O
MW: 99.09 g/mol
Melting Pt: ca 210 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00022384
CAS Number: 6126-22-3
EINECS: 228-095-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR