(Acetylmethylene)triphenylphosphorane 99%

Supplier: Thermo Scientific Chemicals

A11410-09 A11410-18 A11410-30
CAAAA11410-09EA 87.69 CAD
CAAAA11410-09 CAAAA11410-18 CAAAA11410-30
(Acetylmethylene)triphenylphosphorane 99%
(Acetylmethylene)triphenylphosphorane

Fieser: 5,375


Purity: 99% CAS No.: 1439-36-7 Molecular Formula: C21H19OP Formula Weight: 318.36 Storage Temperature: 2°C to 8°C Melting Point: 204-209° Beilstein Reference: 750077 MDL No.: MFCD00008774

Formula: C₂₁H₁₉OP
Melting Pt: 204…209 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00008774
CAS Number: 1439-36-7
EINECS: 215-878-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR