3-(3,4-Dihydroxyphenyl)-L-alanine 98+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Dopar, L-dopa, L-β-3,4-Dihydroxyphenyl-Alanine, 3-Hydroxy-L-tyrosine, Levodopa

A11311-06 A11311-14 A11311-22
CAAAA11311-06EA 73.79 CAD
CAAAA11311-06 CAAAA11311-14 CAAAA11311-22
3-(3,4-Dihydroxyphenyl)-L-alanine 98+%
3-(3,4-Dihydroxyphenyl)-L-alanine

Purity: 98+% CAS No.: 59-92-7 Molecular Formula: C9H11NO4 Formula Weight: 197.19 Odor: Odorless Melting Point: ca 295° dec. Beilstein Reference: 2215169 Merck Reference: 14,5464 MDL No.: MFCD00002598

Formula: C₉H₁₁NO₄
MW: 197.19 g/mol
Melting Pt: ∼ 295 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00002598
CAS Number: 59-92-7
EINECS: 200-445-2
Merck Index: 14,5464

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR