(4-Aminophenyl)acetonitrile 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-(4-Aminophenyl)acetonitrile, 4-Aminobenzyl cyanide

A11304-06 A11304-14
CAAAA11304-06EA 98.28 CAD
CAAAA11304-06 CAAAA11304-14 CAAAA11304-22
(4-Aminophenyl)acetonitrile 99%
(4-Aminophenyl)acetonitrile

Purity: 99% CAS No.: 3544-25-0 Molecular Formula: C8H8N2 Formula Weight: 132.17 Flash Point: 110°(230°F) Melting Point: 42-46° Boiling Point: 312° Beilstein Reference: 1100108 UN No.: 3439 MDL No.: MFCD00007912

Formula: C₈H₈N₂
Boiling Pt: 312 °C
Melting Pt: 42…46 °C
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007912
CAS Number: 3544-25-0
EINECS: 222-587-4
UN: 3439
ADR: "6.1",III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR