L-Alanine ethyl ester hydrochloride ≥98%
Supplier: Thermo Scientific Chemicals
A11271-06
A11271-14
CAAAA11271-06EA
47.56
CAD
CAAAA11271-06
CAAAA11271-14
L-Alanine ethyl ester hydrochloride ≥98%
L-Alanine ethyl ester hydrochloride
Purity: 98+%
CAS No.: 1115-59-9
Molecular Formula: C5H11NO2.HCl
Formula Weight: 153.61
Melting Point: 74-78°
Beilstein Reference: 3594395
Optical Rotation: +2.9° (c=2.5 in water)
MDL No.: MFCD00063662
Formula:
C₅H₁₁NO₂ Melting Pt: 74…78 °C Storage Temperature: Ambient |
MDL Number:
MFCD00063662 CAS Number: 1115-59-9 EINECS: 214-225-9 |
Specification Test Results
Optical rotation | +2.9° (c=2.5 in water) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...