4-Methoxy-3-buten-2-one 90%, tech.
Supplier: Thermo Scientific Chemicals
|
Warning
|
A11235-09
A11235-18
A11235-30
CAAAA11235-09EA
120.4
CAD
CAAAA11235-09
CAAAA11235-18
CAAAA11235-30
4-Methoxy-3-buten-2-one 90%, tech.
4-Methoxy-3-buten-2-one
Purity: 90%
CAS No.: 4652-27-1
Molecular Formula: C5H8O2
Formula Weight: 100.12
Storage Temperature: -30°C to -10°C
Flash Point: 63°(145°F)
Refractive Index: 1.4650
Boiling Point: 66-69°/12mm
Beilstein Reference: 1741053
MDL No.: MFCD00008778
Formula:
C₅H₈O₂ Boiling Pt: 66…69 °C/12 mm Density: 0.990 Flash Pt: 63 °C(145 °F) Storage Temperature: Freezer |
MDL Number:
MFCD00008778 CAS Number: 4652-27-1 EINECS: 225-087-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...