4,4'-Difluorobenzhydryl chloride 98%

Supplier: Thermo Scientific Chemicals
Warning

A11226-06 A11226-09 A11226-18
CAAAAA11226-06EA 56.25 CAD
CAAAAA11226-06 CAAAA11226-09 CAAAA11226-18
4,4'-Difluorobenzhydryl chloride 98%
4,4'-Difluorobenzhydryl chloride

Purity: 98% CAS No.: 27064-94-4 Molecular Formula: C13H9ClF2 Formula Weight: 238.66 Flash Point: 110°(230°F) Refractive Index: 1.5570 Boiling Point: 126-128°/1mm Beilstein Reference: 2052680 UN No.: 1760 MDL No.: MFCD00044329

Formula: C₁₃H₉ClF₂
Boiling Pt: 126…128 °C/1 mm
Density: 1.280
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00044329
CAS Number: 27064-94-4
EINECS: 248-201-4
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR