(1H-Benzoimidazol-2-ylsulfanyl)acetic acid 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A11221-06
A11221-14
CAAAA11221-06EA
81.69
CAD
CAAAA11221-06
CAAAA11221-14
(1H-Benzoimidazol-2-ylsulfanyl)acetic acid 98%
(1H-Benzoimidazol-2-ylsulfanyl)acetic acid
Purity: 98%
CAS No.: 3042-00-0
Molecular Formula: C9H8N2O2S
Formula Weight: 208.24
Melting Point: ca 215° dec.
Beilstein Reference: 11825
MDL No.: MFCD00022673
Formula:
C₉H₈N₂O₂S Melting Pt: ca 215 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00022673 CAS Number: 3042-00-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...