Sodium hexanitritocobaltate(III)
Supplier: Thermo Scientific Chemicals
|
Danger
|
A11216-18
A11216-30
CAAAAA11216-18EA
60.51
CAD
CAAAAA11216-18
CAAAAA11216-30
Sodium hexanitritocobaltate(III)
Sodium hexanitritocobaltate(III)
CAS No.: 14649-73-1
Molecular Formula: CoN6Na3O12
Formula Weight: 403.94
Odor: Odorless
Melting Point: 220° dec.
Merck Reference: 14,8603
MDL No.: MFCD00003512
Formula:
Na₃Co(NO₂)₆ Melting Pt: 220 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00003512 CAS Number: 13600-98-1 EINECS: 237-077-7 UN: 1479 ADR: 5.1,III Merck Index: 14,8603 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...