(±)-1,1'-Bi-2-naphthol 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 1,1'-Binaphthyl-2,2'-Diol, 2,2'-Dihydroxy-1,1'-binaphthyl, 1,1'-Bi-2-naphthol, [1,1'-Binaphthalene]-2,2'-diol

A11182-06 A11182-14 A11182-22
CAAAA11182-06EA 67.15 CAD
CAAAA11182-06 CAAAA11182-14 CAAAA11182-22
(±)-1,1'-Bi-2-naphthol 99%
(±)-1,1'-Bi-2-naphthol

Fieser: 12,190 13,113


Purity: 99% CAS No.: 602-09-5 Molecular Formula: C20H14O2 Formula Weight: 286.33 Odor: Characteristic Melting Point: 215-218° Beilstein Reference: 997518 Merck Reference: 14,1226 UN No.: 2811 MDL No.: MFCD00004068

Formula: C₂₀H₁₄O₂
MW: 286.33 g/mol
Melting Pt: 215…218 °C
Density: 1.301
Storage Temperature: Ambient
MDL Number: MFCD00004068
CAS Number: 602-09-5
EINECS: 210-014-0
UN: 2811
ADR: 6.1 III
Merck Index: 14,1226

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR