Methylcobalamin hydrate 97% (by HPLC)
Supplier: Thermo Scientific Chemicals
A11176-MC
A11176-ME
CAAAA11176-MCEA
152
CAD
CAAAA11176-MC
CAAAA11176-ME
Methylcobalamin hydrate 97% (by HPLC)
Methylcobalamin hydrate
Methylcobalamin hydrate is a coenzyme required in the biosynthesis of methionine.
Vitamin (hematopoietic). It acts as a histamine receptor, Alzheimer research.Methylcobalamin is also used in the treatment of peripheral neuropathy, diabetic neuropathy, and as a preliminary treatment for amyotrophic lateral sclerosis. It can be used to prevent or treat pathology arising from a lack of vitamin B12 such as pernicious anemia.
Formula:
C₆₃H₉₁CoN₁₃O₁₄P·xH₂O Storage Temperature: Freezer |
MDL Number:
MFCD00149221 CAS Number: 288315-09-3 EINECS: 236-535-3 |
Specification Test Results
Appearance (Color) | Red to Dark red to brown |
Form | Powder or crystals |
Assay (HPLC) | ≥96.0% |
Water Content (Karl Fischer Titration) | <12.0% (<10 waters of hydration) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...