1-Methyl-2-pyrrolecarboxaldehyde 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A11163-09
A11163-18
CAAAA11163-09EA
99.07
CAD
CAAAA11163-09
CAAAA11163-18
CAAAA11163-30
1-Methyl-2-pyrrolecarboxaldehyde 98%
1-Methyl-2-pyrrolecarboxaldehyde
Purity: 98%
CAS No.: 1192-58-1
Molecular Formula: C6H7NO
Formula Weight: 109.13
Storage Temperature: 2°C to 8°C
Flash Point: 72°(161°F)
Refractive Index: 1.5610
Boiling Point: 88-90°/22mm
Beilstein Reference: 107811
MDL No.: MFCD00003087
Formula:
C₆H₇NO Boiling Pt: 88…90 °C/22 mm Density: 1.074 Flash Pt: 72 °C(161 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00003087 CAS Number: 1192-58-1 EINECS: 214-755-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...