4-Oxo-4-phenyl-2-butenoic acid predominantly trans 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A11147-06
A11147-14
A11147-22
CAAAA11147-06EA
120.08
CAD
CAAAA11147-06
CAAAA11147-14
CAAAA11147-22
4-Oxo-4-phenyl-2-butenoic acid predominantly trans 97%
4-Oxo-4-phenyl-2-butenoic acid
Purity: 97%
CAS No.: 583-06-2
Molecular Formula: C10H8O3
Formula Weight: 176.17
Odor: Odorless
Melting Point: 93-97°
Beilstein Reference: 1817876
UN No.: 2811
MDL No.: MFCD00014015
Formula:
C₁₀H₈O₃ Melting Pt: 93…97 °C Storage Temperature: Ambient |
MDL Number:
MFCD00014015 CAS Number: 583-06-2 EINECS: 209-496-5 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...