Glycinamide hydrochloride (H-Gly-NH2.HCl) 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A11128-22
A11128-06
A11128-14
CAAAAA11128-22EA
421.86
CAD
CAAAAA11128-22
CAAAA11128-06
CAAAA11128-14
Glycinamide hydrochloride (H-Gly-NH2.HCl) 98%
Glycinamide hydrochloride (H-Gly-NH2.HCl)
Purity: 98%
CAS No.: 1668-10-6
Molecular Formula: NH2C(O)CH2NH2.HCl
Formula Weight: 110.54
Melting Point: ca 204° dec.
Beilstein Reference: 3554199
Formula:
C₂H₇ClN₂O Melting Pt: ca 204 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00013008 CAS Number: 1668-10-6 EINECS: 216-789-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...