α-D-Lactose monohydrate
Supplier: Thermo Scientific Chemicals
A11074-0B
A11074-0I
A11074-30
CAAAA11074-0BEA
107.28
CAD
CAAAA11074-0B
CAAAA11074-0I
CAAAA11074-30
α-D-Lactose monohydrate
α-D-Lactose monohydrate
CAS No.: 5989-81-1
Molecular Formula: C12H22O11.H2O
Formula Weight: 360.32 (342.30anhy)
Odor: Odorless
Melting Point: 207-211° dec.
Beilstein Reference: 3768231
Optical Rotation: +52° (c=4.5 in water, 22h)
MDL No.: MFCD00150747
Formula:
C₁₂H₂₂O₁₁·H₂O Melting Pt: 207…211 °C (dec.) Density: 1.525 Storage Temperature: Ambient |
MDL Number:
MFCD00150747 CAS Number: 5989-81-1 EINECS: 200-559-2 Merck Index: 14,5343 |
Specification Test Results
Optical rotation | +52° (c=4.5 in water, 22h) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...