Isonicotinamidine hydrochloride 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Pyridine-4-carboxamidine hydrochloride, Isonicotinimidamide hydrochloride, 4-Carbamimidoylpyridine hydrochloride, 4-Amidinopyridine hydrochloride, Pyridine-4-carboximidamide hydrochloride, 4-Amidinopyridinium chloride
A11050-06
A11050-14
CAAAA11050-03EA
51.38
CAD
CAAAA11050-03
CAAAA11050-06
CAAAA11050-14
Isonicotinamidine hydrochloride 98+%
Isonicotinamidine hydrochloride
Purity: 98+%
CAS No.: 6345-27-3
Molecular Formula: C6H7N3.HCl
Formula Weight: 157.60
Melting Point: ca 245° dec.
Beilstein Reference: 3562678
Formula:
C₆H₇N₃·ClH MW: 157.601 g/mol Melting Pt: ∼ 245 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00040385 CAS Number: 6345-27-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...