S,S'-Dimethyl-N-cyanodithioiminocarbonate 95%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Dimethyl N-Cyanodithioiminocarbonate, Dimethyl N-Cyanodithiocarbonimidate
A11043-14
A11043-22
A11043-36
CAAAA11043-14EA
60.67
CAD
CAAAA11043-14
CAAAA11043-22
CAAAA11043-36
S,S'-Dimethyl-N-cyanodithioiminocarbonate 95%
S,S'-Dimethyl-N-cyanodithioiminocarbonate
Purity: 95%
CAS No.: 10191-60-3
Molecular Formula: C4H6N2S2
Formula Weight: 146.23
Flash Point:
Formula:
C₄H₆N₂S₂ MW: 146.23 g/mol Melting Pt: 50…56 °C Flash Pt: >110 °C (230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009825 CAS Number: 10191-60-3 EINECS: 233-467-6 UN: 2923 ADR: 8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...