Ethyl-N-hydroxyacetimidate 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Ethyl acetohydroximate

A11006-06 A11006-14 A11006-22
CAAAA11006-06EA 58.3 CAD
CAAAA11006-06 CAAAA11006-14 CAAAA11006-22
Ethyl-N-hydroxyacetimidate 97%
Ethyl N-hydroxyacetimidate

Purity: 97% CAS No.: 10576-12-2 Molecular Formula: C4H9NO2 Formula Weight: 103.12 Storage Temperature: 2°C to 8°C Flash Point: 77°(171°F) Refractive Index: 1.4340 Melting Point: 23-25° Boiling Point: 55-58°/6mm Beilstein Reference: 1700260 MDL No.: MFCD00002114

Formula: C₄H₉NO₂
MW: 103.12 g/mol
Boiling Pt: 55…58 °C (6 mmHg)
Melting Pt: 23…25 °C
Flash Pt: 77 °C (171 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00002114
CAS Number: 10576-12-2
EINECS: 234-165-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR