Triethyl-4-phosphonocrotonate cis- and trans-mixture 94%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A10995-14
A10995-09
CAAAAA10995-14EA
322.32
CAD
CAAAAA10995-14
CAAAA10995-09
CAAAA10995-30
Triethyl-4-phosphonocrotonate cis- and trans-mixture 94%
Triethyl-4-phosphonocrotonate
Purity: 94%
CAS No.: 10236-14-3
Molecular Formula: C10H19O5P
Formula Weight: 250.23
Flash Point:
Formula:
C₁₀H₁₉O₅P Boiling Pt: 133…135 °C/0.4 mm Density: 1.130 Flash Pt: >110 °C(230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009192 CAS Number: 10236-14-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...