4-Bromobenzylamine hydrochloride 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (4-Bromophenyl)methanamine hydrochloride

A10979-09 A10979-18
CAAAA10979-09EA 320.74 CAD
CAAAA10979-09 CAAAA10979-18
4-Bromobenzylamine hydrochloride 98%
4-Bromobenzylamine hydrochloride

Purity: 98% CAS No.: 26177-44-6 Molecular Formula: C7H8BrN.HCl Formula Weight: 222.52 Melting Point: 272-274° Beilstein Reference: 3693458

Formula: C₇H₉BrClN
MW: 222.52 g/mol
Melting Pt: 272…274 °C
Storage Temperature: Ambient
MDL Number: MFCD00012860
CAS Number: 26177-44-6
EINECS: 247-503-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR