2-Cyanopyrazine 97+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Pyrazine-2-carbonitrile, 2-Pyrazinecarbonitril, Pyrazinecarbonitrile

A10974-22 A10974-14
CAAAAA10974-22EA 301.78 CAD
CAAAAA10974-22 CAAAA10974-14
2-Cyanopyrazine 97+%
2-Cyanopyrazine

Purity: 97+% CAS No.: 19847-12-2 Molecular Formula: C5H3N3 Formula Weight: 105.10 Flash Point: 96°(204°F) Refractive Index: 1.5360 Melting Point: 18-20° Boiling Point: 90-93°/15mm Beilstein Reference: 110008 UN No.: 3276 MDL No.: MFCD00049361

Formula: C₅H₃N₃
MW: 105.10 g/mol
Boiling Pt: 90…93 °C/15 mm
Melting Pt: 18…20 °C
Density: 1.174
Flash Pt: 96 °C(204 °F)
Storage Temperature: Ambient
MDL Number: MFCD00049361
CAS Number: 19847-12-2
EINECS: 243-369-5
UN: 3276
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR