N-Carbobenzoxy-L-tyrosine, (max. 10% H₂O) 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Z-Tyr-OH, N-Cbz-L-tyrosine, Cbz-Tyr-OH
A10966-06
A10966-14
CAAAA10966-06EA
57.83
CAD
CAAAA10966-06
CAAAA10966-14
N-Carbobenzoxy-L-tyrosine, (max. 10% H₂O) 99%
N-Carbobenzoxy-L-tyrosine
Purity: 99%
CAS No.: 1164-16-5
Molecular Formula: C17H17NO5
Formula Weight: 315.33
Storage Temperature: 2°C to 8°C
Melting Point: 57-60°
Beilstein Reference: 2169918
Optical Rotation: +10° (c=3 in dioxane)
MDL No.: MFCD00037180
Formula:
C₁₇H₁₇NO₅ MW: 315.33 g/mol Melting Pt: 57…60 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00037180 CAS Number: 1164-16-5 EINECS: 214-609-6 |
Specification Test Results
Optical rotation | +10° (c=3 in dioxane) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...