1-(3-Dimethylaminopropyl)-3-ethylcarbodiimide methiodide 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: EDC Methiodide, 3-(((Ethylimino)methylene)amino)-N,N,N-trimethylpropan-1-aminium iodide, 1-[3-(Dimethylamino)propyl]-3-ethylcarbodiimide Methiodide

A10962-03 A10962-06 A10962-14
CAAAA10962-03EA 45.82 CAD
CAAAA10962-03 CAAAA10962-06 CAAAA10962-14
1-(3-Dimethylaminopropyl)-3-ethylcarbodiimide methiodide 98%
1-(3-Dimethylaminopropyl)-3-ethylcarbodiimide methiodide

Purity: 98% CAS No.: 22572-40-3 Molecular Formula: CH3CH2N=C=N(CH2)3N(CH3)3I Formula Weight: 297.17 Storage Temperature: 2°C to 8°C Melting Point: 90-92° Beilstein Reference: 4162760 MDL No.: MFCD00011833

Formula: CH₃CH₂N=C=N(CH₂)₃N(CH₃)₃I
MW: 297.17 g/mol
Melting Pt: 90…92 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00011833
CAS Number: 22572-40-3
EINECS: 245-095-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR