2,4-Dimethyl-6-hydroxypyrimidine 99%

Supplier: Thermo Scientific Chemicals

Synonyms: 2,6-Dimethylpyrimidin-4-ol, 2,6-Dimethyl-4-pyrimidinol, 6-Hydroxy-2,4-dimethylpyrimidine

A10960-06 A10960-14
CAAAA10960-06EA 44.4 CAD
CAAAA10960-06 CAAAA10960-14
2,4-Dimethyl-6-hydroxypyrimidine 99%
2,4-Dimethyl-6-hydroxypyrimidine

Purity: 99% CAS No.: 6622-92-0 Molecular Formula: C6H8N2O Formula Weight: 124.14 Melting Point: 198-200° Beilstein Reference: 114324 MDL No.: MFCD00006105

Formula: C₆H₈N₂O
MW: 124.14 g/mol
Melting Pt: 198…200 °C
Storage Temperature: Ambient
MDL Number: MFCD00006105
CAS Number: 6622-92-0
EINECS: 229-577-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR