Picolinamidine hydrochloride 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A10942-03
A10942-06
CAAAAA10942-14EA
1730.53
CAD
CAAAAA10942-14
CAAAA10942-03
CAAAA10942-06
Picolinamidine hydrochloride 97%
Picolinamidine hydrochloride
Purity: 97%
CAS No.: 51285-26-8
Molecular Formula: C6H7N3.HCl
Formula Weight: 157.60
Melting Point: 148-152°
Beilstein Reference: 3562671
Formula:
C₆H₈ClN₃ Melting Pt: 148…152 °C Storage Temperature: Ambient |
MDL Number:
MFCD00052271 CAS Number: 51285-26-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...