Tripalmitin (glycerol tripalmitate) 98%
Supplier: Thermo Scientific Chemicals
A10922-03
A10922-06
A10922-14
CAAAA10922-03EA
72.52
CAD
CAAAA10922-03
CAAAA10922-06
CAAAA10922-14
Tripalmitin (glycerol tripalmitate) 98%
Tripalmitin (Glycerol tripalmitate)
Purity: 98%
CAS No.: 555-44-2
Molecular Formula: C51H98O6
Formula Weight: 807.34
Melting Point: 60-64°
MDL No.: MFCD00008995
Formula:
C₅₁H₉₈O₆ Melting Pt: 60…64 °C Storage Temperature: Freezer |
MDL Number:
MFCD00008995 CAS Number: 555-44-2 EINECS: 209-098-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...