Thiophene-2-ethylamine 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 2-(Thiophen-2-yl)ethanamine, 2-(2-Aminoethyl)thiophene

A10920-03 A10920-06 A10920-14
CAAAA10920-03EA 63.67 CAD
CAAAA10920-03 CAAAA10920-06 CAAAA10920-14
Thiophene-2-ethylamine 98%
Thiophene-2-ethylamine

Purity: 98% CAS No.: 30433-91-1 Molecular Formula: C6H9NS Formula Weight: 127.21 Storage Temperature: 2°C to 8°C Flash Point: 87°(188°F) Refractive Index: 1.5510 Boiling Point: 201° Beilstein Reference: 106962 UN No.: 2735 MDL No.: MFCD00051495

Formula: C₆H₉NS
MW: 127.21 g/mol
Boiling Pt: 201 °C
Density: 1.087
Flash Pt: 87 °C (188 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00051495
CAS Number: 30433-91-1
EINECS: 250-196-9
UN: 2735
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR