Methyl-2-(methylthio)benzoate 98%
Supplier: Thermo Scientific Chemicals
A10905-03
A10905-06
CAAAA10905-03EA
73.31
CAD
CAAAA10905-03
CAAAA10905-06
Methyl-2-(methylthio)benzoate 98%
Methyl 2-(methylthio)benzoate
Purity: 98%
CAS No.: 3704-28-7
Molecular Formula: C9H10O2S
Formula Weight: 182.24
Odor: Stench
Melting Point: 64-66°
Beilstein Reference: 2084294
MDL No.: MFCD00068039
Formula:
C₉H₁₀O₂S MW: 182.24 Melting Pt: 64…66 °C Storage Temperature: Ambient |
MDL Number:
MFCD00068039 CAS Number: 3704-28-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...