Methanesulfonamide 98+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Methylsulphonamide, Methylsulfonamide

A10885-09 A10885-18 A10885-30
CAAAAA10885-09EA 60.2 CAD
CAAAAA10885-09 CAAAAA10885-18 CAAAAA10885-30
Methanesulfonamide 98+%
Methanesulphonamide

Purity: 98+% CAS No.: 3144-09-0 Molecular Formula: CH5NO2S Formula Weight: 95.13 Melting Point: 87-93° Beilstein Reference: 1740835

Formula: CH₅NO₂S
MW: 95.13 g/mol
Melting Pt: 87…93 °C
Storage Temperature: Ambient
MDL Number: MFCD00007940
CAS Number: 3144-09-0
EINECS: 221-553-6

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR